Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 50µL Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Axygen Scientific ZT50RS
 View more versions of this product
Catalog No. 14-222-920
  Check Availability
Add to Cart
    Specifications
    Product Type Non-filtered, robotic tip
    Material Polypropylene
    Quantity 96 Tips/Rack
    For Use With (Equipment) Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head, Zymark™/Caliper RapidPlate™/SciClone™ with HV96 head/CAS-4200-1ch.-20-150μL/sec. head
    Sterility Sterile
    Autoclavable Yes
    Tip Style Racked
    Shape Round
    Packaging Rack
    Certifications/Compliance Non-ANSI
    Volume (Metric) 50μL
    Color Clear
    Rack Color Light Green
  • Description & Specifications
Related Products
Product Content Feedback