Learn More
Axygen™ AxyMats™ Sealing Mats for 500uL 96 Well Plates
Catalog No. p-4879395 Shop All Axygen Products
Click to view available options
Pre-slit:
No
Yes
Sterility:
Non-sterile
Sterile
This item is not returnable. View return policy
This item is not returnable. View return policy
Minimize evaporation in wide range of temperatures in PCR and storage applications
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
TRUSTED_SUSTAINABILITY
Specifications
Color White
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
No. per Pack 10 Mats
Pre-slit Yes
Product Type Slitted Microplate Sealing Mat
Plate Compatibility 96-well Deep Well Plate, 0.5 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.