Learn More
Axygen™ AxyPrep Mag™ Tissue Genomic DNA Kits
Shop All Axygen ProductsDescription
The AxyPrep MAG Tissue-Blood gDNA Kit utilizes a magnetic beads-based technology to isolate genomic DNA from rodent tail tissues. This protocol provides instruction to extract gDNA from fresh or frozen rodent tails. The protocol is performed in 96-well format and 1.5 mL tubes. Purification begins by the addition of lysis buffer, and Proteinase K to rupture cell membranes and digest protein. Genomic DNA is then immobilized on magnetic particles by the addition of a magnetic binding reagent. This differential binding allows the gDNA to be easily separated from contaminants using a magnetic field. Contaminants can then be rinsed away using a simple washing procedure.
- Expanded protocol for animal tissue, cultured cells, buccal swabs, and blood
Specifications
Specifications
| Format | 96-well Plate, Tube |
| For Use With (Application) | Isolation of genomic DNA from Rodent tail tissues |
| Includes | 125 mL TB1 Buffer, 120 mL TB2 Buffer, 88 mL Wash Buffer, 10 mL Proteinase K solution, 4.4 mL FB particles, 120 mL Elution Buffer, 1 protocol |
| No. of Reactions | 384 Preps |
| Product Type | Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.