Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AY 9944 dihydrochloride
GSA_VAGREENER_CHOICESDP

Catalog No. 163910 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-391-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 16-391-0 Supplier R&D Systems™ Supplier No. 1639/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of Hedgehog (Hh) signaling. Inhibits Δ7-dehydrocholesterol reductase

Inhibitor of hedgehog (Hh) signaling, possibly via several mechanisms. Inhibits Δ7-dehydrocholesterol reductase (IC50 = 13 nM), thus reduces cholesterol biosynthesis, and also inhibits cholesterol esterification. May also directly block the cellular response to Hh proteins. Teratogenic in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 366-93-8
Molecular Formula C22H30Cl4N2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 464.3
Purity >99%
Molecular Weight (g/mol) 464.296
Chemical Name or Material AY 9944 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.