Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZ 10606120 dihydrochloride
GREENER_CHOICE

Catalog No. 332310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-231-0 10 mg
33-235-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-231-0 Supplier Tocris Bioscience™ Supplier No. 3323/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent P2X7 receptor antagonist

Potent P2X7 receptor antagonist (KD values are 1.4 and 19 nM at human and rat P2X7 receptors respectively). Binds in a positive cooperative manner to sites distinct from, but coupled to, the ATP binding site and acts as a negative allosteric modulator. Inhibits tumor growth and displays antiangiogenic effects in mice.

Chemical Identifiers

CAS 607378-18-7
Molecular Formula C25H36Cl2N4O2

Specifications

Quantity 10 mg
Formula Weight 495.48
Purity >98%
Molecular Weight (g/mol) 495.489
Chemical Name or Material AZ 10606120 dihydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.