Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZ 14145845
SDP

Catalog No. 802350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-235-0 50 mg
80-231-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 80-235-0 Supplier Tocris Bioscience™ Supplier No. 8023/50
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mer/Axl Dual inhibitor

AZ 14145845 is a potent, and highly selective dual Mer/Axl kinase inhibitor (pIC50 = 7 nM and 7.8 nM in pAxl and pMer, respectively). In vivo, AZ 14145845 shows dose-dependent reduction of tumor growth in the Mer and Axl kinase Ba/F3 tumor xenograft model. AZ 14145845 combined with anti-PD1 antibodies and ionizing radiation increases survival in mice with MC38 tumors in vivo. AZ 14145845 is orally bioavailable.

Specifications

Chemical Name or Material 3-[4-[(Dimethylamino)methyl]phenyl]-N-methyl-N-[5-[3-methyl-5-(1,3,5-trimethyl-1H-pyrazol-4-yl)-2-pyridinyl]-1,3,4-oxadiazol-2-yl]imidazo[1,2-a]pyridine-6-methanamine
CAS 2830555-70-7
Quantity 50 mg
Target Additional RTK Inhibitors
Molecular Formula C32H35N9O
Purity 0.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.