Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AZ 20
GREENER_CHOICESDP

Catalog No. 519850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-985-0 50 mg
51-981-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-985-0 Supplier R&D Systems™ Supplier No. 5198/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective ATR kinase inhibitor; antitumor

AZ 20 is a potent and selective ATR kinase inhibitor (IC50 = 5 nM). Exhibits 7.6-fold selectivity over mTOR and selectivity over a panel of 442 kinases, including ATM kinase, PI3-K isoforms, and DNA-PK. Inhibits cell growth in cell lines with high baseline levels of replication stress. Displays antitumor effects in vivo.

Specifications

Quantity 50 mg
Product Line Tocris
Product Type AZ 20
Purity >98%
Molecular Weight (g/mol) 412.51
Inhibitors AZ 20

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.