Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AZ 3146
GREENER_CHOICESDP

Catalog No. 399450 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-945-0 50 mg
39-941-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-945-0 Supplier R&D Systems™ Supplier No. 3994/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective Mps1 kinase inhibitor

Potent and selective monopolar spindle 1 (Mps1) kinase inhibitor (IC50 = 35 nM). Displays selectivity over 46 other kinases including Cdk1 and aurora kinase B. Interferes with chromosome alignment and overrides spindle assembly checkpoint. Inhibits the recruitment of Mad1, Mad2 and centromere protein E (CENP-E) to kinetochores.

Chemical Identifiers

CAS 1124329-14-1
Molecular Formula C24H32N6O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 452.55
Purity >99%
Molecular Weight (g/mol) 452.559
Chemical Name or Material AZ 3146
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.