Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZ 628
GREENER_CHOICE

Catalog No. 483610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-361-0 10 mg
48-365-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-361-0 Supplier Tocris Bioscience™ Supplier No. 4836/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent Raf kinase inhibitor

Potent, ATP-competitive inhibitor of Raf kinases (IC50 values are 29, 34 and 105 nM for c-Raf1, B-RafV600E and wild-type B-Raf, respectively). Displays selectivity for Raf kinases over a panel of 150 other kinases; inhibits activation of tyrosine protein kinases such as VEGFR2, Lyn, Flt1 and Fms. Also inhibits growth, and induces cell cycle arrest and apoptosis in colon and melanoma cell lines with the B-RafV600E mutation.

Chemical Identifiers

CAS 878739-06-1
Molecular Formula C27H25N5O2

Specifications

Quantity 10 mg
Formula Weight 451.52
Purity >98%
Molecular Weight (g/mol) 451.53
Chemical Name or Material AZ 628
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.