Learn More
Description
Specifications
Specifications
| Chemical Name or Material | 3-(2-Methyl-4-pyrimidinyl)-1-(phenylmethyl)-1H-pyrrolo[2,3-c]pyridine |
| CAS | 1679330-37-0 |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in 1eq. HCl and to 100mM in DMSO |
| Molecular Formula | C19H16N4 |
| Formula Weight | 300.36 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

