Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZ PFKFB3 67, >98%
GREENER_CHOICESDP

Catalog No. 574210 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-421-0 10 mg
57-425-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-421-0 Supplier Tocris Bioscience™ Supplier No. 5742/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective PFKFB3 inhibitor

Potent and selective PFKFB3 inhibitor (IC50 values are 11, 159 and 1130 nM for PFKFB3, PFKFB2 and PFKFB1 respectively).

Chemical Identifiers

CAS 1704741-11-6
Molecular Formula C26H25N5O3

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Formula Weight 455.51
Purity >98%
Molecular Weight (g/mol) 455.518
Chemical Name or Material (2S)-N-[4-[[3-Cyano-1-[(3,5-dimethyl-4-isoxazolyl)methyl]-1H-indol-5-yl]oxy]phenyl]-2-pyrrolidinecar
Recommended Storage Store at -29°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.