Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AZ1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10068227
Description
AZ1 Polyclonal specifically detects AZ1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| AZ1 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| 5-azacytidine induced 1, 5-azacytidine-induced protein 1, AZ1, centrosomal protein 131 kDa, centrosomal protein of 131 kDa, Cep131, KIAA1118, pre-acrosome localization protein 1, ZA1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 22994 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 2-5 μg/mg lysate | |
| EAW89632.1 | |
| CEP131 | |
| The immunogen recognized by this antibody maps to a region between residue 1033 and 1083 of human centrosomal protein 131 kDa (5-azacytidine induced 1) using the numbering given in entry EAW89632.1 (GeneID 22994). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction