Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azadibenzocyclooctyne-Amine, PROMO, Thermo Scientific Chemicals

Catalog No. J65637MCPM
Click to view available options
Quantity:
100 mg

Applications

Azadibenzocyclooctyne amine is a carbonyl reactive reagent used to incorporate ADIBO into organic compounds, surfaces or particlesAzadibenzocyclooctyne amine is widely useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

Notes

Heat sensitive. Incompatible with amines and azides.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1255942-06-3
Molecular Formula C18H16N2O
Molecular Weight (g/mol) 276.34 g/mol
MDL Number MFCD22380759
InChI Key OCCYFTDHSHTFER-UHFFFAOYSA-N
Synonym dbco-amine, azadibenzocyclooctyne-amine, adibo-ch2 2-nh2.tfa, azadibenzocyclooctyne-ch2 2-amine.tfa, dibenzocyclooctyne-amine, for copper-free click chemistry, 5-beta-alanyl-11,12-didehydro-5,6-dihydrodibenzo b,f azocine, 3-amino-1-5-aza-3,4:7,8-dibenzocyclooct-1-yne-5-yl-1-propanone, 3-amino-1-2-azatricyclo 10.4.0.0?,? hexadeca-1 16 ,4,6,8,12,14-hexaen-10-yn-2-yl propan-1-one, 3-amino-1-2-azatricyclo 10.4.0.04,9 hexadeca-1 12 ,4 9 ,5,7,13,15-hexaen-10-yn-2-yl propan-1-one
PubChem CID 77078258
SMILES C1C2=CC=CC=C2C#CC3=CC=CC=C3N1C(=O)CCN

Specifications

Quantity 100 mg
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne-amine
TSCA No
Recommended Storage Store at -20°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.