Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azathioprine, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18600823 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-823 100 g Amber Glass Bottle
18-600-820 5 g Amber Glass Bottle
18-600-821 25 g Amber Glass Bottle
18-600-824 1 kg Amber Glass Bottle
18-600-822 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-600-823 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A1383100GM
Add to Cart
Add to Cart

A1383, 446-86-6, C9H7N7O2S

Spectrum™ Chemical Azathioprine, USP is an immunosuppressive drug used in autoimmune diseases and organ transplants. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 446-86-6
Molecular Formula C9H7N7O2S
Molecular Weight (g/mol) 277.26
InChI Key LMEKQMALGUDUQG-UHFFFAOYSA-N
IUPAC Name 6-[(1-methyl-4-nitro-1H-imidazol-5-yl)sulfanyl]-7H-purine
SMILES CN1C=NC(=C1SC1=C2NC=NC2=NC=N1)[N+]([O-])=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98 to 102%
Grade USP
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.