Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZD 1208
GREENER_CHOICESDP

Catalog No. 631050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-105-0 50 mg
63-101-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 63-105-0 Supplier Tocris Bioscience™ Supplier No. 6310/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent pan Pim kinase inhibitor

AZD 1208 is a potent pan Pim kinase inhibitor (IC50 values are 0.4, 1.9 and 5.0 nM for Pim-1, 3 and 2, respectively). Exhibits >43-fold higher affinity for Pim kinases over a range of other kinases. Induces cell cycle arrest and apoptosis in AML cell lines in vitro and inhibits growth of tumor xenografts in vivo. AZD 1208 downmodulates the expression of Mayaro virus (MAYV) structural protein E1 and non-structural protein 1 (nsP1), and it reduces MAYV replication in a dose-dependent manner in HeLa or HDFs cells. Chemosensitizer.

Specifications

Chemical Name or Material (5Z)-5-[[2-[(3R)-3-Amino-1-piperidinyl][1,1'-biphenyl]-3-yl]methylene]-2,4-thiazolidinedione
CAS 1204144-28-4
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C21H21N3O2S
Formula Weight Observed MW: 379.48
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.