Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZD 2461
GREENER_CHOICE

Catalog No. 606010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-601-0 10 mg
60-605-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 60-601-0 Supplier Tocris Bioscience™ Supplier No. 6060/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent PARP inhibitor; orally bioavailable

AZD 2461 is a potent PARP inhibitor (IC50 values are 2, 5 and 200 nM for PARP2, PARP1 and PARP3, respectively). Exhibits anticancer effects in BRCA1 mutant, but not wild-type breast cancer cell lines in vitro. Inhibits growth of olaparib -resistant mammary tumors in a mouse model and is a poor substrate for the P-gp transporter. Orally bioavailable.

Chemical Identifiers

CAS 1174043-16-3
Molecular Formula C22H22FN3O3

Specifications

Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Formula Weight 395.43
Physical Form Solid
Purity >98%
Molecular Weight (g/mol) 395.434
Chemical Name or Material 4-[[4-Fluoro-3-[(4-methoxy-1-piperidinyl)carbonyl]phenyl]methyl]-1(2H)-phthalazinone
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.