Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ AZD 3988
GREENER_CHOICESDP

Catalog No. 483750 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-375-0 50 mg
48-371-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-375-0 Supplier R&D Systems™ Supplier No. 4837/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective DGAT-1 inhibitor

AZD 3988 is a potent and selective diacylglycerol acyltransferase (DGAT-1) inhibitor (IC50 = 0.6 nM). Exhibits selectivity for DGAT-1 over DGAT-2, Kv11.1 (hERG) and cytochrome P450 enzymes. Suppresses triacylglyceride (TAG) plasma excursion and adipose tissue TAG synthesis in rats. Reduces body weight of diet-induced obese rats. Cell permeable and orally bioavailable.

Specifications

Quantity 50 mg
Product Line Tocris
Product Type AZD 3988
Purity >98%
Molecular Weight (g/mol) 456.44
Inhibitors AZD 3988

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.