Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZD 5438
GREENER_CHOICE

Catalog No. 396810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-681-0 10 mg
39-685-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-681-0 Supplier Tocris Bioscience™ Supplier No. 3968/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cdk inhibitor; potently inhibits cdk1, 2, 5 and 9

Potent inhibitor of cyclin dependent kinases (cdks; reported IC50 values are 6 - 45, 14, 16 and 20 nM for cdk2, cdk5, cdk1 and cdk9 respectively). Also inhibits cdk4 and cdk7 in the sub micromolar range. Exhibits antiproliferative activity in human tumor cell lines. Blocks cell cycling at G2-M, S and G1 phases; reduces the proportion of actively cycling cells in vivo.

Chemical Identifiers

CAS 602306-29-6
Molecular Formula C18H21N5O2S

Specifications

Quantity 10 mg
Formula Weight 371.46
Purity >99%
Molecular Weight (g/mol) 371.459
Chemical Name or Material AZD 5438
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.