Learn More
Tocris Bioscience™ AZD 5582 dihydrochloride
Description
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | AZD 5582 dihydrochloride |
| Purity | >97% |
| Molecular Weight (g/mol) | 1088.21 |
| Inhibitors | AZD 5582 dihydrochloride |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.