Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZD 7762 hydrochloride

Catalog No. 519910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-991-0 10 mg
51-995-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-991-0 Supplier Tocris Bioscience™ Supplier No. 5199/10
This item is not returnable. View return policy

Potent and selective ATP-competitive inhibitor of Chk1 and Chk2

AZD 7762 hydrochloride is a potent and selective ATP-competitive inhibitor of Chk1 and Chk2 (IC50 vales are 5 nM for both kinases); displays at least >10 fold selectivity over a panel of 164 kinases. Potentiates cytotoxicity of DNA-damaging agents. Active in vivo. Also improves efficiency of CRISPR-Cpf1-mediated genome editing in hPSC lines (2.7-fold at 1 μM).

Specifications

Quantity 10 mg
Product Type AZD 7762 hydrochloride
Purity >98%
Molecular Weight (g/mol) 398.88
Inhibitors AZD 7762 hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.