Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZD 8055
GREENER_CHOICESDP

Catalog No. 784050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-405-0 50 mg
78-401-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 78-405-0 Supplier Tocris Bioscience™ Supplier No. 7840/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent and selective mTOR inhibitor

AZD 8055 is a highly potent, selective and ATP-competitive mTOR inhibitor (IC50 = 0.8 nM). Inhibits growth and induces autophagy in a range of cancer cell lines and xenografts, including NSCLC and glioblastoma (IC50 values are 20, 50 and 53 nM in H838, A549 and U87MG cell lines, respectively). Orally bioavailable.

Specifications

Chemical Name or Material 5-[2,4-Bis[(3S)-3-methyl-4-morpholinyl]pyrido[2,3-d]pyrimidin-7-yl]-2-methoxybenzenemethanol
CAS 1009298-09-2
Quantity 50 mg
Target mTOR Inhibitors
Molecular Formula C25H31N5O4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.