Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ AZD 9291
SDP

Catalog No. 887350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
88-735-0 50 mg
88-731-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 88-735-0 Supplier Tocris Bioscience™ Supplier No. 8873/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Potent mutant-selective EGFR inhibitor

AZD 9291 is a potent mutant-selective EGFR inhibitor (IC50 values are 1 and 12 nM against EGFR mutants L858R/T790M and L858R, respectively). AZD 9291 exhibited nearly 200 times greater potency against L858R/T790M than wild-type EGFR. In vivo, AZD 9291 causes tumor regression in mutant EGFR xenograft models of non-small cell lung cancer (NSCLC) and transgenic mutant T790M EGFR disease models. AZD 9291 is orally bioavailable.

Specifications

Chemical Name or Material N-[2-[[2-(Dimethylamino)ethyl]methylamino]-4-methoxy-5-[[4-(1-methyl-1H-indol-3-yl)-2-pyrimidinyl]amino]phenyl]-2-propenamide
CAS 1421373-65-0
Quantity 50 mg
Target EGF Receptor Inhibitors
Molecular Formula C28H33N7O2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.