Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azelaic acid, 98%

Catalog No. p-7050252
Click to view available options
Quantity:
25 g
100 g
5 x 100 g

CAS: 123-99-9 | C9H16O4 | 188.223 g/mol

Useful in lubrication and plasticizers. Azelaic acid is used to treat acne.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-99-9
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00004432
InChI Key BDJRBEYXGGNYIS-UHFFFAOYSA-N
Synonym azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110
PubChem CID 2266
ChEBI CHEBI:48131
IUPAC Name nonanedioic acid
SMILES C(CCCC(=O)O)CCCC(=O)O

Specifications

Melting Point ∼102°C
Boiling Point 286°C (100mmHg)
Beilstein 1101094
Merck Index 14,905
Solubility Information Soluble in water
Formula Weight 188.22
Percent Purity 98%
Density 1.225 g/mL
Quantity 25 g
Flash Point 215°C (419°F)
Physical Form Crystalline
Chemical Name or Material Azelaic acid
Show More Show Less
EINECSNumber 204-669-1
RTECSNumber CM1980000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.