Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Azelaic acid dichloride

Catalog No. 29969325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
100 mg

Catalog No. 29-969-325G

Supplier: Sigma Aldrich 29969325G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Please note that sigma-aldrich provides this product to early discovery researchers as part of a collection of unique chemicals
  • Sigma-aldrich does not collect analytical data for this product
  • Buyer assumes responsibility to confirm product identity and/or purity
  • All sales are final.notwithstanding any contrary provision contained in sigma-aldrich's standard terms and conditions of sale or an agreement between sigma-aldrich and buyer, sigma-aldrich sells this product as-is and makes no representation or warranty whatsoever with respect to this product, including any (a) warranty of merchantability; (b) warranty of fitness for a particular purpose; or (c) warranty against infringement of intellectual property rights of a third party; whether arising by law, course of dealing, course of performance, usage of trade or otherwise

Specifications

CAS 123-98-8
Density 1.143 g/mL (at 25°C (literature))
Boiling Point 166°C (18 mmHg)
Molecular Formula C9H14Cl2O2
Linear Formula ClOC(CH2)7 COCl
Refractive Index n20/D 1.467 (literature)
MDL Number MFCD00000767
Quantity 100 mg
Synonym Azelaoyl chloride; Nonanedioyl chloride; Nonanedioyl dichloride
Molecular Weight (g/mol) 225.11
Percent Purity 98%
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.