Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Azer Scientific Formagel™ Tissue Fixation and Transport Gel

Click to view available options
Quantity:
10mL
Description
- Maintains specimen integrity
- Fixation results identical to standard 10% Neutral Buffered Formalin
- Viscoscity enables safe transport without spills
For leak-free transport and fixation of tissue specimens. Compatible with routine, special and IHC staining procedures.
Specifications
Specifications
| Closure Type | Screw-on |
| Color | Clear |
| Content And Storage | 10% Neutral Buffered Formalin |
| Quantity | 10mL |
| For Use With (Application) | Routine, special, IHC staining procedures |
| Material | Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction