Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific Pre-Filled 10% Neutral Buffered Formalin
SDP

Catalog No. 22026441
Change view
Click to view available options
Capacity:
20 mL
40 mL
60 mL
90 mL
120 mL
Content And Storage:
10 mL NBF 10%
20 mL NBF 10%
30 mL NBF 10%
45 mL NBF 10%
60 mL NBF 10%
Packaging Type:
12 Pk., 20 Pks/Cs.
12 Pk., 4 Pks/Cs.
24 Pk., 2 Pks/Cs.
24 Pk., 20 Pks/Cs.
24 Pk., 8 Pks/Cs.
27 Pk., 12 Pks/Cs.
32 Pk., 24 Pks/Cs.
32 Pk., 4 Pks/Cs.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Content And Storage Packaging Type
22-026-441 90 mL 45 mL NBF 10% 12 Pk., 4 Pks/Cs.
22-026-435 20 mL 10 mL NBF 10% 32 Pk., 4 Pks/Cs.
22-026-436 20 mL 10 mL NBF 10% 32 Pk., 24 Pks/Cs.
22-026-437 40 mL 20 mL NBF 10% 24 Pk., 2 Pks/Cs.
22-026-438 40 mL 20 mL NBF 10% 24 Pk., 20 Pks/Cs.
22-026-440 60 mL 30 mL NBF 10% 27 Pk., 12 Pks/Cs.
22-026-442 90 mL 45 mL NBF 10% 12 Pk., 20 Pks/Cs.
22-026-443 120 mL 60 mL NBF 10% 24 Pk., 2 Pks/Cs.
22-026-444 120 mL 60 mL NBF 10% 24 Pk., 8 Pks/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-026-441 Supplier Azer Scientific Supplier No. PFNBF90
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed specifically for the collection and transport of various sizes of tissue samples

  • A traditional fixative that is buffered to a neutral pH
  • Ideal for histological applications for fixing and storing tissue specimens
  • Each container is half filled with 10 percent neutral buffered formalin
  • Packaged with our proprietary Sure-Lok closure

Compliance

Containers are 95 kPa compliant.

Specifications

Capacity 90 mL
Content And Storage 45 mL NBF 10%
Packaging Type 12 Pk., 4 Pks/Cs.
Product Type Formalin
For Use With (Application) Fixative, safe collection, preservation, transportation, and storage of specimens of various sizes
Format Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.