Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Azer Scientific Pre-Filled 10% Neutral Buffered Formalin

Click to view available options
Capacity:
20 mL
40 mL
60 mL
90 mL
120 mL
Content And Storage:
10 mL NBF 10%
20 mL NBF 10%
30 mL NBF 10%
45 mL NBF 10%
60 mL NBF 10%
Packaging Type:
12 Pk., 20 Pks/Cs.
12 Pk., 4 Pks/Cs.
24 Pk., 2 Pks/Cs.
24 Pk., 20 Pks/Cs.
24 Pk., 8 Pks/Cs.
27 Pk., 12 Pks/Cs.
32 Pk., 24 Pks/Cs.
32 Pk., 4 Pks/Cs.
Description
- A traditional fixative that is buffered to a neutral pH
- Ideal for histological applications for fixing and storing tissue specimens
- Each container is half filled with 10 percent neutral buffered formalin
- Packaged with our proprietary Sure-Lok closure
Compliance
Containers are 95 kPa compliant.
Specifications
Specifications
Capacity | 120 mL |
Content And Storage | 60 mL NBF 10% |
Packaging Type | 24 Pk., 8 Pks/Cs. |
Product Type | Formalin |
For Use With (Application) | Fixative, safe collection, preservation, transportation, and storage of specimens of various sizes |
Format | Container |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction