Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AZGP1 Monoclonal Antibody (21), Invitrogen™
Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
Antigen | AZGP1 |
---|---|
Clone | 21 |
Concentration | 1.59 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543490
|
Invitrogen™
MA543490 |
50 μL |
Each for $285.50
|
|
|||||
PIMA543038
|
Invitrogen™
MA543038 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human Alpha-2-glycoprotein/AZGP1
AZGP1 stimulates lipid degradation in adipocytes and causes the extensive fat losses associated with some advanced cancers. It may bind polyunsaturated fatty acids.Specifications
AZGP1 | |
1.59 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative | |
alpha-2 - glycoprotein 1 zinc; alpha-2 - glycoprotein 1, zinc; alpha-2-glycoprotein 1, zinc; alpha-2-glycoprotein 1, zinc-binding; Alpha-2-glycoprotein, zinc; Azgp1; major histocompatibility complex class I-like protein; testicular tissue protein Li 227; ZA2G; ZA2GA; Zag; Zinc a 2 glycoprotein; Zinc a2glycoprotein; Zinc alpha2glycoprotein; zinc alpha-2-glycoprotein 1; Zinc a 2 glycoprotein; Zinc a2glycoprotein; zinc-alpha-2-glycoprotein; Zn - alpha2 - glycoprotein; Zn a 2 glycoprotein; Zn a 2 GP; Zn alpha 2 glycoprotein; Zn alpha 2 GP; Zn a 2 glycoprotein; Zn a 2 GP; Zna2gp; zn-alpha-2-glycoprotein; Zn-alpha2-glycoprotein; zn-alpha-2-GP; ZNGP1 | |
AZGP1 | |
IgG1 | |
Protein A | |
Antibody |
21 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
P25311 | |
563 | |
Recombinant Human Alpha-2-glycoprotein/AZGP1 Protein. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
AZGP1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title