Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AZGP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104403 Supplier Invitrogen™ Supplier No. PA5104403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AZGP1.

Alpha-2-glycoprotein 1, zinc-binding (AZGP1) is a gene encoding a protein that belongs to the immunoglobulin superfamily. This protein is abundant in various body fluids, such as blood plasma and milk, and is involved in lipid mobilization. AZGP1 is expressed in numerous tissues, including the liver, prostate, and adipose tissue, where it plays a significant role in the regulation of lipid metabolism and adipocyte function. It acts as an adipokine, influencing the breakdown and mobilization of fats, thereby contributing to energy homeostasis. In cancer research, AZGP1 has been highlighted for its potential as a prognostic biomarker, particularly in prostate and breast cancer, where lower expression levels are often associated with poor prognosis. Recent studies have also linked AZGP1 to cachexia, an extreme form of weight loss and muscle wasting, by promoting lipid mobilization in adipose tissue and influencing systemic catabolic states. Due to its multifaceted roles in lipid metabolism and disease processes, AZGP1 is considered a promising target for therapeutic interventions in metabolic and cancer-related disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AZGP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene AZGP1
Gene Accession No. P25311, Q64726
Gene Alias alpha-2 - glycoprotein 1 zinc; alpha-2 - glycoprotein 1, zinc; alpha-2-glycoprotein 1, zinc; alpha-2-glycoprotein 1, zinc-binding; Alpha-2-glycoprotein, zinc; Azgp1; major histocompatibility complex class I-like protein; testicular tissue protein Li 227; ZA2G; ZA2GA; Zag; Zinc a 2 glycoprotein; Zinc a2glycoprotein; Zinc alpha2glycoprotein; zinc alpha-2-glycoprotein 1; Zinc a 2 glycoprotein; Zinc a2glycoprotein; zinc-alpha-2-glycoprotein; Zn - alpha2 - glycoprotein; Zn a 2 glycoprotein; Zn a 2 GP; Zn alpha 2 glycoprotein; Zn alpha 2 GP; Zn a 2 glycoprotein; Zn a 2 GP; Zna2gp; zn-alpha-2-glycoprotein; Zn-alpha2-glycoprotein; zn-alpha-2-GP; ZNGP1
Gene Symbols AZGP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human AZGP1(Accession P25311), corresponding to amino acid residues K102-P152.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12007, 563
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.