Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AZI1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982513
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2513 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2513 Supplier Bioss Supplier No. BS12977R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

AZI1, also named as CEP131 and KIAA1118, may play a role in spermatogenesis. It is located at the ciliary base and required for cilium formation.

Specifications

Antigen AZI1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CEP131
Gene Accession No. Q9UPN4
Gene Alias 5-azacytidine induced 1; 5-azacytidine induced gene 1; 5-azacytidine-induced protein 1; Az1; Azi; Azi1; centrosomal protein 131; centrosomal protein 131 kDa; centrosomal protein 131kDa; Centrosomal protein of 131 kDa; Cep131; KIAA1118; Pre-acrosome localization protein 1; ZA1
Gene Symbols CEP131
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human AZI1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22994
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.