Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AZIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109436 Supplier Invitrogen™ Supplier No. PA5109436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKKVVENDKF PSGVEKTGSD EPAFMYYMND GVYGSFASKL SEDLNTIPEV HKKYKEDEPL FTSSLWGPSC DELDQIVESC LLPELNVGDW LIFDNMGADS FHEPSAFNDF QRPAIYYMMS FSDWYEMQDA GITSDSMMKN FFFVPSCIQL SQEDSFSAEA.

Antizyme inhibitor (AZIN1) protein that positively regulates ornithine decarboxylase (ODC) activity and polyamine uptake. AZI is an enzymatically inactive ODC homolog that counteracts the negative effect of ODC antizymes (AZs) OAZ1, OAZ2 and OAZ3 on ODC activity by competing with ODC for antizyme-binding. Inhibits antizyme-dependent ODC degradation and releases ODC monomers from their inactive complex with antizymes, leading to formation of the catalytically active ODC homodimer and restoring polyamine production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AZIN1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene AZIN1
Gene Accession No. O14977, O35484, Q63764
Gene Alias 1700085L02Rik; antizyme inhibitor 1; AZI; AZIA1; AZIN1; O OAZIN; OAZI; Oazin; ODC antizyme inhibitor; ODC1L; Ornithine decarboxylase antizyme inhibitor
Gene Symbols AZIN1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 289-448 of human AZIN1 (NP_056962.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51582, 54375, 58961
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.