Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azodicarbonamide, 97%

Catalog No. p-61733
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic bottle

CAS: 123-77-3 | C2H4N4O2 | 116.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X Bond Formation (Halogen)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-77-3
Molecular Weight (g/mol) 116.08
MDL Number MFCD00007958
InChI Key XOZUGNYVDXMRKW-AATRIKPKSA-N
Synonym azodicarbonamide, azodicarboxamide, diazenedicarboxamide, azobisformamide, azodiformamide, azodicarbamide, azodicarboamide, azobiscarbonamide, azobiscarboxamide, celogen az
PubChem CID 5462814
IUPAC Name (E)-carbamoyliminourea
SMILES C(=O)(N)N=NC(=O)N

Specifications

Melting Point 195.0°C to 202.0°C
Assay Percent Range 97%
Packaging Plastic bottle
Molecular Formula C2H4N4O2
Linear Formula H2NCON=NCONH2
Quantity 2.5 kg
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in common solvents, soluble in dimethyl sulfoxide, slightly soluble in demethylformamide
Formula Weight 116.08
Percent Purity 97%
Chemical Name or Material Azodicarbonamide, 97%
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-650-8
RTECSNumber LQ1040000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.