Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Azoxystrobin Monoclonal Antibody (4B7)
GREENER_CHOICE

Catalog No. PIMA549735
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549735 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549735 Supplier Invitrogen™ Supplier No. MA549735
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

Azoxystrobin is a fungicide that belongs to the class of strobilurin fungicides. It is widely used in agriculture to protect crops from fungal diseases such as powdery mildew, leaf spots, and rusts. Azoxystrobin works by inhibiting the respiratory chain of fungal cells, disrupting their energy production and leading to their death. It is also used in the treatment of plant diseases caused by soil-borne pathogens. Azoxystrobin has a broad spectrum of activity against various fungal species and is effective in controlling both foliar and soil-borne diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Azoxystrobin
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 4B7
Concentration 13 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of AXS hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.