Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aztreonam

Catalog No. aaj6288703
Click to view available options
Quantity:
1 g
5 g

CAS: 78110-38-0 | C13H17N5O8S2 | 435.426 g/mol

Aztreonam is used to treat severe infections of the urinary tract, lower respiratory tract, skin, stomach, female reproductive organs, and other body systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aztreonam is used to treat severe infections of the urinary tract, lower respiratory tract, skin, stomach, female reproductive organs, and other body systems.

Solubility
Soluble in DMF/water (1:1) at 50 mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78110-38-0
Molecular Formula C13H17N5O8S2
Molecular Weight (g/mol) 435.426
MDL Number MFCD00072145
InChI Key WZPBZJONDBGPKJ-PSGLRMFWSA-N
PubChem CID 56842327
IUPAC Name 2-[(E)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(2R,3R)-2-methyl-4-oxo-1-sulfoazetidin-3-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoic acid
SMILES CC1C(C(=O)N1S(=O)(=O)O)NC(=O)C(=NOC(C)(C)C(=O)O)C2=CSC(=N2)N

Specifications

Type Aztreonam
Physical Form Powder
Melting Point 227°C (decomposition)
Color White
Quantity 1 g
Merck Index 14,925
Solubility Information Soluble in DMF/water (1:1) at 50 mg/ml
Formula Weight 435.43
Chemical Name or Material Aztreonam
EINECSNumber 278-839-9
RTECSNumber UA2451400
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.