Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aztreonam, G-Biosciences
SCI_ED

Catalog No. S000040
Click to view available options
Quantity:
1 g
5 g

Aztreonam is a monobactam Beta-lactam antibiotic whose structure contains only a single ring. Its unique ring structure makes it resistant to Beta-lactamases.

Aztreonam is effective against gram-negative bacteria and against P. aeruginosa.

Chemical Identifiers

Molecular Weight (g/mol) 435.43 g/mol

Specifications

Quantity 1 g
Percent Purity >95%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.