Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azure I

Catalog No. aaa1750806
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 531-55-5 | C15H16ClN3S | 305.824 g/mol

Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie's modified Nocht's method for paraffin sections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie′s modified Nocht′s method for paraffin sections.

Solubility
Soluble in 1 mg/ml water. Sparingly soluble in alcohol.

Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 531-55-5
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) 305.824
MDL Number MFCD00011935
InChI Key DNDJEIWCTMMZBX-UHFFFAOYSA-N
Synonym azure b, azure i, methylene azure b, 3-dimethylamino-7-methylamino phenothiazin-5-ium chloride, trimethylthionine chloride, azur i, methyleneazure, methylene azure van, unii-7c8bel8wqv, modr kresylova bb czech
PubChem CID 68275
IUPAC Name dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride
SMILES CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]

Specifications

Melting Point ∼205°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 3922630
Merck Index 14,6058
Solubility Information Soluble in 1mg/ml water. Sparingly soluble in alcohol.
Formula Weight 305.83
Chemical Name or Material Azure I
Hazard Category Warning
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 208-511-2
RTECSNumber SO5550000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.