Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Azure Biosystems AzureSpectra Fluorescent Western Blot Kit with Protein-Free Block

Kit contains materials for 10 fluorescent Western blots, including LF PVDF membranes, wash buffer, protein-free blocking buffer, and secondary antibodies.
$329.50
Specifications
Product Type | Western Blot Kit |
---|---|
Detection Method | Fluorescence |
For Use With (Application) | Western Blot |
Membrane Compatibility | Nitrocellulose, PVDF |
Quantity | 10 Mini Blots |
Catalog Number | Mfr. No. | Kit Contents | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Kit Contents | Price | Quantity & Availability | |||||
AC2104AZ
![]() |
Azure Biosystems
AC2104 |
Includes: Secondary Ab rb650/ms550 |
Each for $329.50
|
|
|||||
AC2140AZ
![]() |
Azure Biosystems
AC2140 |
Includes: Secondary Ab rb700/ms800 |
Each for $329.50
|
|
|||||
AC2141AZ
![]() |
Azure Biosystems
AC2141 |
Includes: Secondary Ab ms700/rb800 |
Each for $329.50
|
|
|||||
Description
- Full kits for two-color fluorescent Western blots. Also offered with Fluorescent Block.
- Each kit contains enough materials for 10 Western blots:
- 9 x 7 cm Low Fluorescence PVDF Membranes, 10 membranes
- 10 x Fluorescent Blot Washing Buffer, 250 mL
- 5 x Protein-Free Blot Blocking Buffer, 60 mL
- 2 x Background Quenching Sheets
- 2 x 40 μL Fluorescent Secondary Antibodies
Specifications
Western Blot Kit | |
Western Blot | |
10 Mini Blots |
Fluorescence | |
Nitrocellulose, PVDF |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title