Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Azurocidin/CAP37/HBP Antibody (308), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289593AF532
Description
Azurocidin/CAP37/HBP Monoclonal antibody specifically detects Azurocidin/CAP37/HBP in Human samples. It is validated for ELISASpecifications
Azurocidin/CAP37/HBP | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
308 | |
ELISA | |
AZAMP, AZU, azurocidin, azurocidin 1, CAP37Heparin-binding protein, cationic antimicrobial protein 37, HBPCationic antimicrobial protein CAP37, HUMAZUR, NAZC, neutrophil azurocidin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Azurocidin/CAP37/HBP (NP_001691.1; Met1-Pro250). | |
0.1 mL | |
Apoptosis | |
566 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction