Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Azurocidin/CAP37/HBP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$231.00 - $262.00
Specifications
Antigen | Azurocidin/CAP37/HBP |
---|---|
Dilution | ELISA 1:1000-1:2000 |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Azurocidin/CAP37/HBP Polyclonal antibody specifically detects Azurocidin/CAP37/HBP in Human samples. It is validated for ELISASpecifications
Azurocidin/CAP37/HBP | |
ELISA | |
Unconjugated | |
Rabbit | |
Apoptosis | |
0.2 um filtered solution in PBS | |
566 | |
IgG | |
Antigen and protein A Affinity-purified |
ELISA 1:1000-1:2000 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
AZAMP, AZU, azurocidin, azurocidin 1, CAP37Heparin-binding protein, cationic antimicrobial protein 37, HBPCationic antimicrobial protein CAP37, HUMAZUR, NAZC, neutrophil azurocidin | |
Produced in rabbits immunized with purified, recombinant Human Azurocidin/CAP37/HBP (Accession#: NP_001691.1; Met 1-Pro 250). AZU1 specific IgG was purified by Human AZU1 affinity chromatography . | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title