Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ B-7569 Vinyl GHS L3A Diamond Labels for BBP85 Printer
Allows printer to create 2-color GHS/CLP chemical labels in one print pass
$182.00 - $273.00
Specifications
Description | Red on White |
---|---|
Adhesive Type | Acrylic |
Color | Red, White |
Material | Vinyl (B-7569) |
Product Type | GHS Diamond Label |
Catalog Number | Mfr. No. | dimDimensionsLxW | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | dimDimensionsLxW | Quantity | Price | Quantity & Availability | ||||
19-000-568
|
Brady™
130774 |
70/Pk. |
Each for $273.00
|
|
|||||
19-000-120
|
Brady™
130768 |
90/Pk. |
Each for $182.00
|
|
|||||
Description
- White labels with pre-printed diamonds
- Indoor/outdoor grade withstands exposure to moisture, chemicals and handling
- Available in L1, L2, L3A, L3B, L4A and L4B diamond legends
Specifications
Red on White | |
Red, White | |
GHS Diamond Label | |
BBP™85 Printer |
Acrylic | |
Vinyl (B-7569) | |
L3A Diamond + Text |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title