Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-Chlorocatecholborane 96.0+%, TCI America™
SDP

Catalog No. C166925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C166925G 25 g
C16695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C166925G Supplier TCI America Supplier No. C166925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55718-76-8
Molecular Formula C6H4BClO2
Molecular Weight (g/mol) 154.36
MDL Number MFCD00043395
InChI Key AZYGEWXDKHFOKB-UHFFFAOYSA-N
Synonym 2-Chloro-1,3,2-benzodioxaborole
PubChem CID 4136924
IUPAC Name 2-chloro-2H-1,3,2-benzodioxaborole
SMILES ClB1OC2=CC=CC=C2O1

Specifications

Melting Point 59°C
Quantity 25 g
UN Number 2925
Formula Weight 154.36
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material B-Chlorocatecholborane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.