Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ B-PER™ Bacterial Protein Extraction Reagent

Catalog No. PI90084
Click to view available options
Quantity:
165 mL
250 mL
500 mL

Efficiently lyse E.coli and other bacterial cells and gently extract native proteins with these nonionic detergent solutions.

Perform efficient, gentle lysis and extraction of E.coli and other bacterial cells with Thermo Scientific™ B-PER Bacterial Protein Extraction Reagents.

These easy-to-use cell lysis reagents are nonionic detergent solutions that effectively disrupt cells and solubilize native or recombinant proteins without denaturation. Certain formulations include DNase I and lysozyme enzyme supplements to improve yield of large molecular weight proteins that are difficult to purify. B-PER Reagents are compatible with the addition of standard protease inhibitors and with downstream protein purification methods, such as GST purification and His-tagged immobilized metal affinity chromatography (IMAC).

Highlights:

  • Ready to use – B-PER Reagents provide one-step E.coli cell lysis by a mild, nonionic detergent (proprietary) in your choice of Tris or phosphate buffer formulations
  • Fast and simple – just add B-PER Reagent to a bacterial pellet, shake for 10 minutes and recover soluble proteins after pelleting the cell debris
  • Excellent yields – recover both soluble and insoluble recombinant protein from bacterial lysates and purify inclusion bodies to near-homogeneous levels
  • Flexible – B-PER Reagents are suitable for any scale of protein extraction and are available in phosphate and 1X and 2X Tris formulations, with and without enzymes
  • Compatible – completely compatible with addition of protease inhibitor cocktails, and resulting protein extract can be used in protein assays, typical affinity purification methods (e.g., GST, 6xHis) and other applications

Convenient, ready-to-use formats:

  • B-PER and B-PER II (2X) Reagents are free of enzymatic components
  • B-PER Reagent (in phosphate buffer) is an amine-free formulation for direct compatibility with labeling and crosslinking
  • B-PER with Enzymes and B-PER Direct with Enzymes include lysozyme and DNase I for improved lysis and recovery

Includes:

Reagent packages are available in Tris and phosphate buffer formulations; Kits contain Tris-based B-PER Reagent plus lysozyme and DNase I enzymes.

TRUSTED_SUSTAINABILITY

Specifications

Description B-PER Bacterial Protein Extraction Reagent
Quantity 250 mL
Product Type Bacterial Protein Extraction Reagent
Volume (Metric) 250 mL
Content And Storage Store B-PER Reagent at ambient temperature. Store Lysozyme and DNaseI at -20°C.
Product Line B-PER™
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.