Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ B-PER™ Complete Bacterial Protein Extraction Reagent, 500 mL PROMO
Click to view available options
:
Thermo Scientific™ B-PER™ Complete Bacterial Protein Extraction Reagent
Description
- An easy-to-use cell lysis reagent that effectively disrupts cells and solubilizes native or recombinant proteins without denaturation
- All B-PER Reagents are compatible with downstream applications such as affinity chromatography (e.g., immobilized metal affinity chromatography, glutathione chromatography), SDS-PAGE, and protein quantification (e.g., Pierce BCA Protein Assay, Pierce 660nm Protein Assay)
- This;all-inclusive formulation combines the lysis reagent with lysozyme, and a universal nuclease to enable mild extraction from both gram-negative and gram-positive bacteria
- Depending on the particular application protease inhibitors, salts, reducing agents, denaturants, and chelating agents may be added to the reagent.
Features of the B-PER Bacterial Protein Extraction Reagent:
- Detergent in Tris buffer with Lysozyme and Universal Nuclease
- Improved cell lysis and DNA digestion, thereby releasing soluble proteins and reducing viscosity to increase yields
- Recovery of large molecular weight proteins (high MW proteins)
- Cost-effective equivalent to Merck Millipore Bugbuster™ Master Mix
- More effective than traditional sonication and typical homemade lysis buffers, many of which include detergents and components that interfere with downstream applications
Specifications
Specifications
| Formulation | Proprietary detergent in 20mM Tris buffer, pH 7.5, containing lysozyme and universal nuclease |
| Quantity | 500 mL |
| Product Type | Bacterial Protein Extraction Reagent |
| Volume (Metric) | 500 mL |
| Content And Storage | Store at 4°C. |
| Product Line | B-PER |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction