Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ B-pure Deionization Systems: Cartridges
SCI_ED

Catalog No. p-2371710
Click to view available options
Capacity:
1725 grains as NaCl, 875
760 grains as NaCl, 875
Type:
Demineralizer Mult Cartridge
Type M Ultrapure Cartridge
This item is not returnable. View return policy
This item is not returnable. View return policy

Accessory cartridges for deionization systems.

  • Type M Ultrapure for use with B-Pure Deionization System
  • Resin type: Mixed bed
  • High purity
TRUSTED_SUSTAINABILITY

Specifications

Capacity 760 grains as NaCl, 875
Resin Type Mixed Bed
For Use With (Equipment) B-Pure™ deionization system and analytical lab testing
Purity High
Type Type M Ultrapure Cartridge
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.