Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ B-Raf (K601E Mutant) Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143383
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
This gene encodes a protein belonging to the RAF family of serine/threonine protein kinases This protein plays a role in regulating the MAP kinase/ERK signaling pathway, which affects cell division, differentiation, and secretion Mutations in this gene, most commonly the V600E mutation, are the most frequently identified cancer-causing mutations in melanoma, and have been identified in various other cancers as well, including non-Hodgkin lymphoma, colorectal cancer, thyroid carcinoma, non-small cell lung carcinoma, hairy cell leukemia and adenocarcinoma of lung Mutations in this gene are also associated with cardiofaciocutaneous, Noonan, and Costello syndromes, which exhibit overlapping phenotypes A pseudogene of this gene has been identified on the X chromosome.
Specifications
| B-Raf (K601E Mutant) | |
| Polyclonal | |
| Unconjugated | |
| P29466, P48729, Q8N752 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 122011, 1452, 834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.54 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% Proclin 300; pH 7 | |
| B-raf; B-Raf proto-oncogene, serine/threonine kinase; BRAF1; B-RAF1; NS7; RAFB1 | |
| Carrier-protein conjugated synthetic peptide surrounding mutant K601E of human B-Raf. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction