Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B-Raf Mouse anti-Human, Clone: IHC600, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP3005171ML
Description
B-Raf Monoclonal specifically detects B-Raf in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
B-Raf | |
Monoclonal | |
Unconjugated | |
Tris Buffer, pH 7.3 - 7.7, with 1% BSA | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
IHC600 | |
Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
94 kDa B-raf protein, B-Raf proto-oncogene serine/threonine-protein kinase (p94), B-RAF1, BRAF1MGC126806, EC 2.7.11.1, MGC138284, murine sarcoma viral (v-raf) oncogene homolog B1, NS7, p94, Proto-oncogene B-Raf, RAFB1, serine/threonine-protein kinase B-raf, v-raf murine sarcoma viral oncogene homolog B1FLJ95109 | |
A peptide covering the V600E mutation. | |
1 mL | |
Apoptosis, Phospho Specific | |
673 | |
Store at 4°C. Do not freeze. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction