Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ B-Raf Recombinant Rabbit Monoclonal Antibody (6G3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549885
Description
B-Raf Recombinant Monoclonal Antibody for IHC (P), ELISA
BRAF is involved in the transduction of mitogenic signals from the cell membrane to the nucleus. It is activated by somatic point mutation in human cancer, and is thought to play a role in the postsynaptic responses of hippocampal neuron. This protein plays a role in regulating the MAP kinase/ERKs signaling pathway, which affects cell division, differentiation, and secretion. Mutations in B-Raf have also been associated with various cancers, including non-Hodgkin lymphoma, colorectal cancer, malignant melanoma, thyroid carcinoma, non-small cell lung carcinoma, and adenocarcinoma of lung. A pseudogene, which is located on chromosome X, has been identified for B-Raf.
Specifications
B-Raf | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P15056 | |
BRAF | |
A synthesized peptide derived from human B Raf. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
6G3 | |
Unconjugated | |
BRAF | |
94 kDa B-raf protein; 9930012E13Rik; AA120551; AA387315; AA473386; Braf; B-raf; B-raf protein; B-raf protein isoform 1; B-raf protein isoform 2; B-Raf proto-oncogene serine/threonine-protein kinase; B-Raf proto-oncogene serine/threonine-protein kinase (p94); B-Raf proto-oncogene, serine/threonine kinase; Braf transforming gene; BRAF1; B-RAF1; Braf2; Braf-2; C230098H17; C87398; C-RMIL; D6Ertd631e; FLJ95109; MGC126806; murine sarcoma viral (v-raf) oncogene homolog B1; NS7; p94; Proto-oncogene B-Raf; proto-oncogene c-Rmil; RAFB1; RMIL; rmil serine/threonine-protein kinase; serine/threonine kinase; serine/threonine-protein kinase B-raf; Serine/threonine-protein kinase Rmil; v-raf murine sarcoma viral oncogene homolog B; v-raf murine sarcoma viral oncogene homolog B1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
673 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction