Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-tag Rabbit, Polyclonal, GenScript™

Catalog No. 50272880
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-272-880 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-272-880 Supplier Genscript Corporation Supplier No. A0063840
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Well-characterized antibodies for epitope tags consisting of short sequences have been widely used in the study of protein expression in various systems.The B-tag is an epitope tag composed of a six-residue peptide, QYPALT, corresponding to a highly conserved region of VP7 core protein of the blue tongue virus. The GenScript Rabbit Anti-B-tag Polyclonal Antibody is developed in rabbit hosts using a synthetic B-tag peptide conjugated to KLH.GenScript Rabbit Anti-B-tag Polyclonal Antibody is highly purified from rabbit antiserum by immunoaffinity chromatography and is supplied as a 40 μg aliquot at a concentration of 1 mg/mL.

B-tag Polyclonal antibody specifically detects B-tag in Tag samples. It is validated for ELISA, Western Blot

Specifications

Antigen B-tag
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Alias VP7
Host Species Rabbit
Immunogen B epitope tag peptide QYPALT conjugated to KLH
Purification Method Affinity chromatography
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.