Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ B1 Bradykinin Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518988
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518988 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518988 Supplier Invitrogen™ Supplier No. PA518988
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.

Bradykinin, a 9 aa peptide, is generated in pathophysiologic conditions such as inflammation, trauma, burns, shock, and allergy. Two types of G-protein coupled receptors have been found which bind bradykinin and mediate responses to these pathophysiologic conditions. The protein encoded by this gene is one of these receptors and is synthesized de novo following tissue injury. Receptor binding leads to an increase in the cytosolic calcium ion concentration, ultimately resulting in chronic and acute inflammatory responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen B1 Bradykinin Receptor
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Bdkrb1
Gene Accession No. P46663
Gene Alias B1 bradykinin receptor; B1BKR; B1R; Bdkrb; BDKRB1; BDKRB2; Bk b1 receptor; BK-1 receptor; BKB1R; BKR; BKR1; BKRB1; BRADYB1; bradykinin B1 receptor; bradykinin receptor 1; bradykinin receptor B1; bradykinin receptor B2; bradykinin receptor, beta 1; KB1; kinin B1; kinin B1 receptor
Gene Symbols Bdkrb1
Host Species Goat
Immunogen Synthetic peptide sequence (KVWELYKQCTPK) corresponding to the internal amino acids of BDKRB1 (aa 322-334).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 623
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.