Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
B1 Bradykinin Receptor Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572564
Description
Bradykinin, a 9 aa peptide, is generated in pathophysiologic conditions such as inflammation, trauma, burns, shock, and allergy. Two types of G-protein coupled receptors have been found which bind bradykinin and mediate responses to these pathophysiologic conditions. The protein encoded by this gene is one of these receptors and is synthesized de novo following tissue injury. Receptor binding leads to an increase in the cytosolic calcium ion concentration, ultimately resulting in chronic and acute inflammatory responses.
Specifications
B1 Bradykinin Receptor | |
Polyclonal | |
Unconjugated | |
BDKRB1 | |
BDKRB1; BKB1R; Bradykinin receptor 1; B1BKR; B1R; Bk b1 receptor; BK-1 receptor; BRADYB1; Bradykinin receptor B1; Kinin b1 receptor; B1 bradykinin receptor; BKR1; Bradykinin B1 receptor | |
Rabbit | |
Protein A | |
RUO | |
623 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide | |
P46663 | |
BDKRB1 | |
KLH conjugated synthetic peptide between 213-239 amino acids from the Central region of human BDK_1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction